Learn More
Greiner Bio-One 384-well Small Volume™ HiBase Polystyrene Microplates
GREENER_CHOICE
Catalog No. 07000040 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
White
Sterility:
Non-sterile
Sterile
No. per Case:
40
150
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility No. per Case
07-000-040 Black Non-sterile 150
07-000-041 White Non-sterile 150
07-000-065 Black Non-sterile 40
07-000-066 White Non-sterile 40
07-000-063 Black Sterile 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 07-000-040 Supplier Greiner Bio-One Supplier No. 78407625
Add to Cart
Add to Cart
Features round wells with a conical geometry. Greiner Bio-One 384-Well Small Volume™ HiBase Polystyrene Microplates is free of detectable DNase, RNase, human DNA.
  • Non-Pyrogenic
  • Material: Polystyrene
  • Free of detectable DNase, RNase, human DNA
  • Traced way back to production through defined LOT number system
  • Working volume: 4μL - 25μL
Certifications
DIN ISO 9001
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Description Plate design: HiBase
Array 16 x 24
Binding Type Medium Binding
Color Black
Material Polystyrene
No. of Wells 384
Sterility Non-sterile
No. per Case 150