Learn More
Bellco Glass, Inc. 384 Well Tissue Culture Plate
SDP
Catalog No. 502329857
Encompass_Preferred
Tissue Culture Treated surface improves cell adhesionPlates are labeled with alpha-numeric markings for easy identification of individual wellsLids have a stacking feature for added stability when handling tall stacks of plates384 Well Plates are ideal for automated applications, colorimetric assays, compound storage or small volume applications Pack DescriptionIndividualCase Quantity50SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneTissue Culture TreatedYesBottom ShapeFlatCell Growth Area0.1135cm²RNase/DNase FreeYes
Catalog No. 50-232-9857 Supplier Bellco Glass, Inc. Supplier No. 229138000
Tissue Culture Treated surface improves cell adhesionPlates are labeled with alpha-numeric markings for easy identification of individual wellsLids have a stacking feature for added stability when handling tall stacks of plates384 Well Plates are ideal for automated applications, colorimetric assays, compound storage or small volume applications Pack DescriptionIndividualCase Quantity50SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneTissue Culture TreatedYesBottom ShapeFlatCell Growth Area0.1135cm²RNase/DNase FreeYes