Learn More
Electron Microscopy Sciences NUNC™ 96 Well Optical Bottom Plate White
SDP
Catalog No. 5033295
Encompass_Preferred
The NUNC Coverglass Optical Bottom 96 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well. Minimum light scatter and low autofluorescence. The plate's upper structure is offered in black (for fluorometric detection). Sterile. External dimensions 128 x 86 mm.
Color: White
Catalog No. 50-332-95 Supplier Electron Microscopy Sciences Supplier No. 6481615
The NUNC Coverglass Optical Bottom 96 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well. Minimum light scatter and low autofluorescence. The plate's upper structure is offered in black (for fluorometric detection). Sterile. External dimensions 128 x 86 mm.
Color: White