Learn More
Alkali Scientific ASI™ 384 White Well, Clear Bottom, Cell Culture Plate, Unibinding, Sterile, 10/Cs, PS/ PS
SDP
Catalog No. 503028927
Encompass_Preferred
384-Well Cell Culture Plates: Our high-quality 384 HT Plates are designed for high-throughput research applications. These plates have been successfully tested for fluorescence and luminescence assays, providing high signal detection with low background noise. The alphanumeric labeling ensures easy identification of wells. The total well volume is 120 µL, with a working volume of 100 µL. These plates offer maximum reflection and minimal cross-talk, ideal for luminescence assays. Made from sterile polystyrene, the surface is positively charged and cell culture-treated, making it suitable for adherent cells. Key features: Maximum reflection and minimal cross-talk for luminescence assays HTS compatible Flat-bottom wells Suitable for colorimetry, fluorescence, and luminescence assays Transparent lid included with each well plate 1 plate and lid per inner pack, 10 plates/lids per case
Catalog No. 50-302-8927 Supplier Alkali Scientific Supplier No. TPW3084
Add to Cart
Add to Cart
384-Well Cell Culture Plates: Our high-quality 384 HT Plates are designed for high-throughput research applications. These plates have been successfully tested for fluorescence and luminescence assays, providing high signal detection with low background noise. The alphanumeric labeling ensures easy identification of wells. The total well volume is 120 µL, with a working volume of 100 µL. These plates offer maximum reflection and minimal cross-talk, ideal for luminescence assays. Made from sterile polystyrene, the surface is positively charged and cell culture-treated, making it suitable for adherent cells. Key features: Maximum reflection and minimal cross-talk for luminescence assays HTS compatible Flat-bottom wells Suitable for colorimetry, fluorescence, and luminescence assays Transparent lid included with each well plate 1 plate and lid per inner pack, 10 plates/lids per case