Learn More
Aurora Microplates 384LV WELL BLK/SB NS/NT 40/CS
Catalog No. 50245355
Encompass
Black, Round 384 IQ-LV, Solid bottom, Non-Treated, Non Sterile, Black Notchless Lid, Std. Bar Code, Standard Pack, 40 Plates Per Case
Catalog No. 50-245-355 Supplier Aurora Microplates Supplier No. ABA010100A
Black, Round 384 IQ-LV, Solid bottom, Non-Treated, Non Sterile, Black Notchless Lid, Std. Bar Code, Standard Pack, 40 Plates Per Case