Learn More
Aurora Microplates 384LV200 BLK/CLR NS/NT 40/CS
Catalog No. 50245403
Encompass
Black, Round 384 IQ-LV, 188u, Non-Treated, Non Sterile, Black Notchless Lid, Std. Bar Code, Standard Pack, 40 Plates Per Case
Catalog No. 50-245-403 Supplier Aurora Microplates Supplier No. ABA210100A
Black, Round 384 IQ-LV, 188u, Non-Treated, Non Sterile, Black Notchless Lid, Std. Bar Code, Standard Pack, 40 Plates Per Case