Learn More
Beckman Coulter Microplate, 384-Well, Echo Qualified, Clear, Round Bottom, Sterile, Polypropylene (6 CS)
Catalog No. NC2007959
Encompass
384-well polystyrene microplate with rounded wells and clear bottom for high-throughput screening, parallel reactions, and optical detection in life science research and clinical diagnostics.
Catalog No. NC2007959 Supplier Beckman Coulter Supplier No. C74290
384-well polystyrene microplate with rounded wells and clear bottom for high-throughput screening, parallel reactions, and optical detection in life science research and clinical diagnostics.