Learn More
PERMAGEN LABWARE LTD 384W LOW ELUTION POST PLATE
Catalog No. NC3822555
Encompass_Preferred
384-Well Low Elution Post Magnet Plate with Integrated Cushion Base.The Permagen 384-well Low Elution post magnet plate was designed for use in manual or automation applications for high throughput PCR. Magnetic beads are pulled midway up onto well wall to help eliminate accidental bead pellet disruption and to achieve lower volumes. Includes integrated cushion base for maximum recovery during automation
Catalog No. NC3822555 Supplier PERMAGEN LABWARE LTD Supplier No. MSP384LE
Add to Cart
Add to Cart
384-Well Low Elution Post Magnet Plate with Integrated Cushion Base.The Permagen 384-well Low Elution post magnet plate was designed for use in manual or automation applications for high throughput PCR. Magnetic beads are pulled midway up onto well wall to help eliminate accidental bead pellet disruption and to achieve lower volumes. Includes integrated cushion base for maximum recovery during automation