Learn More
Biotang Inc 384-well PCR plate. Natural. 100/case. Compatible with most robotics, designed to SBS standards. Rigid skirt for gripping by robotic plate handlers. Compatible with ABI 3730 Automated Sequencers- single cut corner!
Catalog No. 507515020 Shop All BIOTANG Inc Products
Encompass_Preferred
384-well PCR plate. Natural. 100/case. Compatible with most robotics, designed to SBS standards. Rigid skirt for gripping by robotic plate handlers. Compatible with ABI 3730 Automated Sequencers- single cut corner!
Catalog No. 50-751-5020 Supplier Biotang Inc Supplier No. BT39690
Add to Cart
Add to Cart
384-well PCR plate. Natural. 100/case. Compatible with most robotics, designed to SBS standards. Rigid skirt for gripping by robotic plate handlers. Compatible with ABI 3730 Automated Sequencers- single cut corner!