Learn More
USA Scientific Inc 384-wl v-bot pp plt,20/slv,nat
Catalog No. 502745478
Encompass
384-well PlateOne conical bottom polypropylene plate, 20/sleeve, 80/case, natural, str
Catalog No. 50-274-5478 Supplier USA Scientific Inc Supplier No. 18238410
384-well PlateOne conical bottom polypropylene plate, 20/sleeve, 80/case, natural, str