Learn More
O Berk Company 38- 400 WHITE RIB SIDE MATTE TOP PLASTIC PP CAP - F217 LINER 3100/Pk
Catalog No. NC9234157
Encompass_Preferred
Size 99.99; Neck Diameter 38.00; Neck Finish 400; Material; PP PLASTIC; LinerF217;Surface Finish RIB SIDE / MATTE TOP 3100/Pk
Catalog No. NC9234157 Supplier O Berk Company Supplier No. 07010037
Size 99.99; Neck Diameter 38.00; Neck Finish 400; Material; PP PLASTIC; LinerF217;Surface Finish RIB SIDE / MATTE TOP 3100/Pk