Learn More
Ace Glass, Inc. PTFE multi-paddle agitator, 38mm diameter
SDP
Catalog No. 509558265
Encompass_Preferred
38mm diameter PTFE multi-paddle agitator for 10mm OD drilled or knob type shafts. Paddles are replaceable.
Catalog No. 50-955-8265 Supplier Ace Glass, Inc. Supplier No. 808904
38mm diameter PTFE multi-paddle agitator for 10mm OD drilled or knob type shafts. Paddles are replaceable.