Learn More
Beckman Coulter Beckman Coulter Titanium Tube Spacer, 38 mm Diameter, for Ultracentrifuge Rotors
Catalog No. NC2741462
Encompass
Beckman Coulter titanium tube spacer (P/N 342697) is a single-piece titanium spacer designed to position and support 38 x 102 mm centrifuge tubes in compatible ultracentrifuge rotors, ensuring correct tube seating and spacing for high-speed operation.
  • 38 mm nominal diameter.
  • Compatible with 38 x 102 mm tubes with a 94 mL maximum fill volume.
  • Made from titanium for strength and corrosion resistance.
  • Designed to ensure proper tube seating and spacing in ultracentrifuge rotors.
  • Supports operation up to 45,000 rpm (≈235,000 x g) when used with compatible tubes.
  • Sold individually.
Catalog No. NC2741462 Supplier Beckman Coulter Supplier No. 342697
Beckman Coulter titanium tube spacer (P/N 342697) is a single-piece titanium spacer designed to position and support 38 x 102 mm centrifuge tubes in compatible ultracentrifuge rotors, ensuring correct tube seating and spacing for high-speed operation.
  • 38 mm nominal diameter.
  • Compatible with 38 x 102 mm tubes with a 94 mL maximum fill volume.
  • Made from titanium for strength and corrosion resistance.
  • Designed to ensure proper tube seating and spacing in ultracentrifuge rotors.
  • Supports operation up to 45,000 rpm (≈235,000 x g) when used with compatible tubes.
  • Sold individually.