Learn More
MSC Three M Heavy Duty Duct Tape, 1.89 in, 8.6 mil
Catalog No. 501113887
Encompass
This heavy-duty duct tape features a rubber adhesive and polyethylene film over cloth scrim backing, ideal for MRO and construction applications. It provides a strong, water-resistant seal, conforms to irregular surfaces, and resists wear and abrasion. The tape holds reliably for up to one year indoors and removes cleanly from indoor opaque, non-glass surfaces for up to one month.
  • Designed for heavy-duty MRO and construction applications
  • Clean removal from indoor opaque, non-glass surfaces for up to one month
  • Reliable rubber adhesive bonds immediately and holds for up to one year indoors
  • Strong polyethylene backing resists water, wear, and abrasion
  • Forms a tight, moisture-resistant seal in various containment situations
  • Conforms to irregular surfaces without lifting for easy application
This item is not returnable. View return policy
Catalog No. 50-111-3887 Supplier MSC Supplier No. 65364721
This heavy-duty duct tape features a rubber adhesive and polyethylene film over cloth scrim backing, ideal for MRO and construction applications. It provides a strong, water-resistant seal, conforms to irregular surfaces, and resists wear and abrasion. The tape holds reliably for up to one year indoors and removes cleanly from indoor opaque, non-glass surfaces for up to one month.
  • Designed for heavy-duty MRO and construction applications
  • Clean removal from indoor opaque, non-glass surfaces for up to one month
  • Reliable rubber adhesive bonds immediately and holds for up to one year indoors
  • Strong polyethylene backing resists water, wear, and abrasion
  • Forms a tight, moisture-resistant seal in various containment situations
  • Conforms to irregular surfaces without lifting for easy application