Learn More
Sigma Aldrich Fine Chemicals Biosciences 395 nm Light source1EA
Catalog No. 502908743
Encompass
The 395nm LED Light Module is used with the Penn PhD Photoreactor M2 for chemists and researchers to accelerate chemical reactions using photoredox catalysis.
Catalog No. 50-290-8743 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7440781EA
The 395nm LED Light Module is used with the Penn PhD Photoreactor M2 for chemists and researchers to accelerate chemical reactions using photoredox catalysis.