Learn More
Prolume Ltd NanoFuel Solvent 1 x 10ml
Catalog No. NC0596803
Encompass
NanoFuel Solvent is NanoLight’s solution to the problem associated with the solubility of Coelenterazine and the many new Coelenterazine analogs that have been made available in recent years. Native Coelenterazine can be dissolved in a 10 times higher concentration compared to other alcohols. NanoFuel-Solvent also guarantees Coelenterazine stability if store at -20°C for a few weeks or at -80°C for up to one year.
Catalog No. NC0596803 Supplier Prolume Ltd Supplier No. 39910
NanoFuel Solvent is NanoLight’s solution to the problem associated with the solubility of Coelenterazine and the many new Coelenterazine analogs that have been made available in recent years. Native Coelenterazine can be dissolved in a 10 times higher concentration compared to other alcohols. NanoFuel-Solvent also guarantees Coelenterazine stability if store at -20°C for a few weeks or at -80°C for up to one year.