Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3A-Amino-3A-deoxy-(2AS,3AS)-beta-cyclodextrin Hydrate 97.0+%, TCI America™
SDP

Catalog No. A19161G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A19161G Supplier TCI America Supplier No. A19161G

Chemical Identifiers

CAS 117194-77-1
Molecular Formula C42H71NO34
Molecular Weight (g/mol) 1134.003
InChI Key OZLVLKFBPJMRCM-FUFZOPRGSA-N
PubChem CID 11320754
SMILES C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O2)C(C8O)O)CO)CO)CO)CO)CO)CO)O)N)O

Specifications

Color White
Quantity 1 g
Formula Weight 1134.00
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3A-Amino-3A-deoxy-(2AS,3AS)-beta-cyclodextrin Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.