Learn More
Medchemexpress LLC 3a-bis-(4-fluorophenyl) methoxytropane (hydrochloride) | 202646-03-5 | 100.0% | 379.87 | C21H24ClF2NO | 25 MG
SDP
Catalog No. 5000213053
Encompass_Preferred
AHN 1-055 hydrochloride is the hydrochloride salt of 3α-bis-(4-fluorophenyl) methoxytropane, a research compound that inhibits dopamine uptake with a reported IC50 of about 71 nM. Supplied as a high-purity solid, it is intended for pharmacological and neurochemical studies of dopamine transporter activity in vitro and in vivo.
  • High-purity solid suitable for research use.
  • Potent dopamine transporter inhibitor (IC50 ≈71 nM).
  • Appropriate for in vitro and in vivo pharmacology studies.
  • Available in small, accurately weighed quantities for dose-ranging.
  • Store sealed at 4°C; in solution store at -80°C for long-term preservation.
Catalog No. 50-002-13053 Supplier Medchemexpress LLC Supplier No. HY10131525MG
AHN 1-055 hydrochloride is the hydrochloride salt of 3α-bis-(4-fluorophenyl) methoxytropane, a research compound that inhibits dopamine uptake with a reported IC50 of about 71 nM. Supplied as a high-purity solid, it is intended for pharmacological and neurochemical studies of dopamine transporter activity in vitro and in vivo.
  • High-purity solid suitable for research use.
  • Potent dopamine transporter inhibitor (IC50 ≈71 nM).
  • Appropriate for in vitro and in vivo pharmacology studies.
  • Available in small, accurately weighed quantities for dose-ranging.
  • Store sealed at 4°C; in solution store at -80°C for long-term preservation.