Learn More
Medchemexpress LLC 3a-hydroxy-6-methyl-1-phenyl-2,3-dihydropyrrolo[2,3-b]quinolin-4-one | 674289-55-5 | MFCD04113775 | 98.9%, 99.6% | C18H16N2O2 | 10MG
SDP
Catalog No. 5000258701
Encompass_Preferred
Blebbistatin is a selective inhibitor of non-muscle myosin II ATPase activity used as a research reagent to probe myosin-dependent contractility, cell migration, cytokinesis, and cytoskeletal dynamics. It is provided as a solid with reported high purity, and manufacturer guidance for storage and solubility for both in vitro and in vivo use.
  • Selective inhibitor of non-muscle myosin II ATPase activity
  • Useful for studying cell migration, contractility, and cytokinesis
  • Supplied as a stable solid with long-term frozen storage
  • High reported purity suitable for research applications
  • DMSO soluble for stock solutions; protocols available for in vivo suspension
Catalog No. 50-002-58701 Supplier Medchemexpress LLC Supplier No. HY1381310MG
Blebbistatin is a selective inhibitor of non-muscle myosin II ATPase activity used as a research reagent to probe myosin-dependent contractility, cell migration, cytokinesis, and cytoskeletal dynamics. It is provided as a solid with reported high purity, and manufacturer guidance for storage and solubility for both in vitro and in vivo use.
  • Selective inhibitor of non-muscle myosin II ATPase activity
  • Useful for studying cell migration, contractility, and cytokinesis
  • Supplied as a stable solid with long-term frozen storage
  • High reported purity suitable for research applications
  • DMSO soluble for stock solutions; protocols available for in vivo suspension