Learn More
Research Products International Corp 3a70B Complete Counting Cocktail, 4 Liters
SDP
Catalog No. 50476433
Encompass_Preferred
High performance counting efficiency with medium to high volumes of aqueous solutions. Optimized blend gives superior performance in the "gel" range. Holds up to 60% aqueous sample. Also recommended for counting insoluble solids and suspensions. Accommodates most aqueous solutions without tedious sample preparation. Highly resistant to quenching.
Catalog No. 50-476-433 Supplier Research Products International Corp Supplier No. 111154
High performance counting efficiency with medium to high volumes of aqueous solutions. Optimized blend gives superior performance in the "gel" range. Holds up to 60% aqueous sample. Also recommended for counting insoluble solids and suspensions. Accommodates most aqueous solutions without tedious sample preparation. Highly resistant to quenching.