Learn More
Medchemexpress LLC 3aH-indole-3a-carboxylic acid, 1-([1,1'-biphenyl]-4-ylmethyl)-hexahydro-3-substituted, sodium salt | 2070009-45-7 | 98.7% | 440.47 g/mol | C25H25N2NaO4 | 10 MG
SDP
Catalog No. 5000215663
Encompass_Preferred
Fumarate hydratase-IN-2 sodium salt is a cell-permeable, competitive inhibitor of fumarate hydratase (Ki = 4.5 μM). Supplied as the sodium salt as a white to off-white solid, it is intended for biochemical and cell-based research, including studies of metabolic regulation and nutrient-dependent cytotoxicity.
  • Cell-permeable competitive inhibitor of fumarate hydratase (Ki = 4.5 μM).
  • Sodium salt form for improved solubility in aqueous and polar solvents.
  • High purity (98.7%) appropriate for research use.
  • Soluble in DMSO (10.66 mg/mL) and water (6.25 mg/mL) with warming and sonication.
  • Stable when stored sealed away from moisture; follow solvent storage recommendations.
Catalog No. 50-002-15663 Supplier Medchemexpress LLC Supplier No. HY100005A10MG
Fumarate hydratase-IN-2 sodium salt is a cell-permeable, competitive inhibitor of fumarate hydratase (Ki = 4.5 μM). Supplied as the sodium salt as a white to off-white solid, it is intended for biochemical and cell-based research, including studies of metabolic regulation and nutrient-dependent cytotoxicity.
  • Cell-permeable competitive inhibitor of fumarate hydratase (Ki = 4.5 μM).
  • Sodium salt form for improved solubility in aqueous and polar solvents.
  • High purity (98.7%) appropriate for research use.
  • Soluble in DMSO (10.66 mg/mL) and water (6.25 mg/mL) with warming and sonication.
  • Stable when stored sealed away from moisture; follow solvent storage recommendations.