Learn More
Medchemexpress LLC 3aH-indole-3a-carboxylic acid, biphenyl-4-ylmethyl derivative, sodium salt | 2070009-45-7 | 98.7% | 440.47 g/mol | C25H25N2NaO4 | 25 MG
SDP
Catalog No. 5000215575
Encompass_Preferred
Fumarate hydratase-IN-2 sodium salt is a cell-permeable, competitive fumarate hydratase inhibitor (Ki = 4.5 μM) that exhibits nutrient-dependent cytotoxicity. Supplied as a white to off-white solid intended for research use, it is suitable for cell-based and biochemical studies of fumarate hydratase activity.
  • Cell-permeable inhibitor effective in cell-based assays.
  • Competitive inhibition with Ki = 4.5 μM for target engagement.
  • High purity (98.7%) suitable for research applications.
  • Soluble in DMSO and water with warming and sonication.
  • Available in small solid quantities and solution formats for flexibility.
Catalog No. 50-002-15575 Supplier Medchemexpress LLC Supplier No. HY100005A25MG
Fumarate hydratase-IN-2 sodium salt is a cell-permeable, competitive fumarate hydratase inhibitor (Ki = 4.5 μM) that exhibits nutrient-dependent cytotoxicity. Supplied as a white to off-white solid intended for research use, it is suitable for cell-based and biochemical studies of fumarate hydratase activity.
  • Cell-permeable inhibitor effective in cell-based assays.
  • Competitive inhibition with Ki = 4.5 μM for target engagement.
  • High purity (98.7%) suitable for research applications.
  • Soluble in DMSO and water with warming and sonication.
  • Available in small solid quantities and solution formats for flexibility.