Learn More
Cayman Chemical 3amino Benzamide
SDP
Supplier:  Cayman Chemical 103975 G
Encompass_Preferred
A PARP inhibitor (IC50 = 5.4 µM in C3H/10T1/2 mouse stem cells); inhibits spontaneous, as well as FGF2-induced, cell invasiveness of HUVECs at 50 µM; induces tubulogenesis and reduces FGF2-induced expression of UPA in HUVECs at 50 µM; reduces telomere length in HeLaS3 human cervical cancer cells and non-cancerous COM3 hamster cells
Catalog No. 50-289-0594