Learn More
Cayman Chemical 3'-Amino-3'-deoxyadenosine-5'-O-diphosphate (sodium salt), Purity: ≥98%, 2.1 mg
SDP
Catalog No. 502915897
Encompass_Preferred
An amine-modified nucleotide; has been used in the synthesis of fluorescently labeled ADP; has been used in the synthesis of phosphorylated probes for GTP pyrophosphokinase domains
Catalog No. 50-291-5897 Supplier Cayman Chemical Supplier No. 383632.1 MG
An amine-modified nucleotide; has been used in the synthesis of fluorescently labeled ADP; has been used in the synthesis of phosphorylated probes for GTP pyrophosphokinase domains