Learn More
Cayman Chemical 3AmIno3deoxyadenosIn5 2.5mg
SDP
Catalog No. 502917486
Encompass_Preferred
An amine-modified nucleotide; has been used as a probe for quantifying ATP substrate specificity for kinases and ligases; terminates RNA strand synthesis catalyzed by RNA polymerase in E. coli
Catalog No. 50-291-7486 Supplier Cayman Chemical Supplier No. 383642.5 MG
An amine-modified nucleotide; has been used as a probe for quantifying ATP substrate specificity for kinases and ligases; terminates RNA strand synthesis catalyzed by RNA polymerase in E. coli