Learn More
Cayman Chemical 3-Amino-5-hydroxybenzoic Acid, Purity: ≥98%, CAS: 76045-71-1, 500 mg
SDP
Catalog No. 502911320
Encompass_Preferred
A biosynthetic precursor to ansamycin antibiotics
Catalog No. 50-291-1320 Supplier Cayman Chemical Supplier No. 35831500 MG
A biosynthetic precursor to ansamycin antibiotics