Learn More
Cayman Chemical 3-amino-L-Tyrosine (hydrochloride), Purity: ≥95%, CAS: 23279-22-3, 100 mg
SDP
Catalog No. 502912947
Encompass_Preferred
A tyrosine derivative; completely inhibits the growth of a tyrosine-sensitive mutant of N. crassa at 400 µM; cytotoxic to KG-1 leukemia cells at 200-400 µg/ml; dietary administration decreases fecundity and hatchability of eggs of B. oleae at 0.01 g/100 ml, as well as survival of adult B. oleae at 6 g/100 ml
Catalog No. 50-291-2947 Supplier Cayman Chemical Supplier No. 36310100 MG
A tyrosine derivative; completely inhibits the growth of a tyrosine-sensitive mutant of N. crassa at 400 µM; cytotoxic to KG-1 leukemia cells at 200-400 µg/ml; dietary administration decreases fecundity and hatchability of eggs of B. oleae at 0.01 g/100 ml, as well as survival of adult B. oleae at 6 g/100 ml