Learn More
Cayman Chemical 3-amino-L-Tyrosine (hydrochloride), Purity: ≥95%, CAS: 23279-22-3, 50 mg
SDP
Numéro de catalogue. 502912598
Encompass_Preferred
A tyrosine derivative; completely inhibits the growth of a tyrosine-sensitive mutant of N. crassa at 400 µM; cytotoxic to KG-1 leukemia cells at 200-400 µg/ml; dietary administration decreases fecundity and hatchability of eggs of B. oleae at 0.01 g/100 ml, as well as survival of adult B. oleae at 6 g/100 ml
Numéro de catalogue. 50-291-2598 Fournisseur Cayman Chemical Code fournisseur 3631050 MG
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
A tyrosine derivative; completely inhibits the growth of a tyrosine-sensitive mutant of N. crassa at 400 µM; cytotoxic to KG-1 leukemia cells at 200-400 µg/ml; dietary administration decreases fecundity and hatchability of eggs of B. oleae at 0.01 g/100 ml, as well as survival of adult B. oleae at 6 g/100 ml
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.