Learn More
Cayman Chemical 3-Aminopropylphosphonic Acid, Purity: ≥95%, CAS: 13138-33-5, 250 mg
SDP
Catalog No. 502994610
Encompass_Preferred
A partial agonist of GABAB receptors (IC50 = 1.5 μM in a radioligand binding assay); induces relaxation in unstimulated isolated guinea pig ileum longitudinal muscle; completely inhibits GABA- and baclofen-induced inhibition of vagally stimulated bronchospasms in guinea pigs (5 mg/kg); reverses the antitussive effect of baclofen in cats (3 mg/kg)
Catalog No. 50-299-4610 Supplier Cayman Chemical Supplier No. 23556250 MG
A partial agonist of GABAB receptors (IC50 = 1.5 μM in a radioligand binding assay); induces relaxation in unstimulated isolated guinea pig ileum longitudinal muscle; completely inhibits GABA- and baclofen-induced inhibition of vagally stimulated bronchospasms in guinea pigs (5 mg/kg); reverses the antitussive effect of baclofen in cats (3 mg/kg)