Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3Ar,4S,5R,6As)-(-)-Hexahydro-5-Hydroxy-4-(Hydroxymethyl)-2H-Cyclopenta[B]Furan-2-One

Catalog No. 3415761G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-157-61G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-157-61G Supplier Sigma Aldrich Supplier No. 3415761G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(3Ar,4S,5R,6As)-(-)-Hexahydro-5-Hydroxy-4-(Hydroxymethyl)-2H-Cyclopenta[B]Furan-2-One

Specifications

CAS 35661-40-6
Melting Point 180°C to 187°C
Molecular Formula C24H21NO4
Linear Formula C24H21NO4
MDL Number MFCD00037128
Quantity 100 g
Molecular Weight (g/mol) 387.43 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.