Learn More
Medchemexpress LLC 3aS 4S 6aR -Biotin- 50mg
SDP
Catalog No. 5000205121
Encompass_Preferred
(3aS 4S 6aR)-Biotin-PEG4-Alkyne is an alkyne-activated biotinylation reagent (3aS 4S 6aR)-Biotin-PEG4-Alkyne can be reacted with azides via a copper-catalyzed click reaction Biotin-labeled biomolecules can be bound to avidin or streptavidin for further purification and detection[1]
Catalog No. 50-002-05121 Supplier Medchemexpress LLC Supplier No. HYW11014650MG
(3aS 4S 6aR)-Biotin-PEG4-Alkyne is an alkyne-activated biotinylation reagent (3aS 4S 6aR)-Biotin-PEG4-Alkyne can be reacted with azides via a copper-catalyzed click reaction Biotin-labeled biomolecules can be bound to avidin or streptavidin for further purification and detection[1]