Learn More
Cayman Chemical 3-azido-7-hydroxy Coumarin, Purity: ≥95%, CAS: 817638-68-9, 100 mg
SDP
Catalog No. 502889718
Encompass_Preferred
A click-reactive fluorescent dye; Upon CuAAC in a biological system, a fluorescent product is produced that displays excitation/emission maxima of 404 and 477 nM, respectively; has been used for spatially restricted metabolite labeling and the detection of foodborne pathogens
Catalog No. 50-288-9718 Supplier Cayman Chemical Supplier No. 10596100 MG
A click-reactive fluorescent dye; Upon CuAAC in a biological system, a fluorescent product is produced that displays excitation/emission maxima of 404 and 477 nM, respectively; has been used for spatially restricted metabolite labeling and the detection of foodborne pathogens