Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Adapter for BNC-4mm Jacks
SDPSCI_ED

Catalog No. S90373 Shop All 3B Scientific Products
Change view
Click to view available options
For Use With (Equipment):
BNC 4mm jacks
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
S90373 BNC 4mm jacks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S90373 Supplier 3B Scientific™ Supplier No. U11259
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adapter for crossover from a BNC plug to 4-mm-jacks with 19 mm spacing.

Adapter for crossover from BNC plug to 4mm jacks.

Specifications

Product Type Adapter
For Use With (Equipment) BNC 4mm jacks
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.