Learn More
3B Scientific™ Airway Management Trainer with Stand
SDPSCI_ED
Catalog No. S07966 Shop All 3B Scientific Products
Change view
Click to view available options
Product Type:
Child Airway Trainer
Infant Airway Trainer
Catalog No. Product Type
S07966 Child Airway Trainer
S07965 Infant Airway Trainer
2 options
Catalog No. S07966
Supplier: 3B Scientific™ W44666
Includes
Stand, pump spray lubricant and hard carrying case
Ideal for practicing airway management skills.
  • Features realistic size and anatomy
  • Realistic anatomy includes mouth, tongue, oral pharynx, larynx, epiglottis, vocal cords, trachea, and esophagus
  • Separate lungs for auscultation
  • Inflatable stomach bladder indicated esophageal insufflation
  • Practice suctioning and oral and nasal intubation techniques
  • With stand to mount head
Order Info
Infant Airway Trainer (S07965) can be attached to a Resusci Baby (W19517) brand CPR manikin.
Specifications
Product Type Child Airway Trainer
For Use With (Application) Practicing Airway Management Skills
Includes Stand, pump spray lubricant and hard carrying case