Learn More
3B Scientific™ Alligator Clip
SDPSCI_ED
Catalog No. S90374 Shop All 3B Scientific Products
Change view
Click to view available options
Includes:
Pk. of 15
Catalog No. Includes
S90374 Pk. of 15
1 options
Catalog No. S90374 Supplier 3B Scientific™ Supplier No. U13805PK
Add to Cart
Add to Cart
Accepts 4mm test leads or any other 4mm Multilam plug.
  • Not insulated With 4mm sockets Connection also possible with screw clamp or soldering
Order Info
Sold in packs of 15
Specifications
Includes Pk. of 15
Product Type Alligator Clip, uninsulated