Learn More
3B Scientific™ Auscultation Trainer and Smartscope Complete Package with Amplifiers
SDPSCI_ED
Catalog No. S07957 Shop All 3B Scientific Products
Change view
Click to view available options
Auscultation Trainer package with Smartscope and Amplifiers
Catalog No.
S07957 Auscultation Trainer package with Smartscope and Amplifiers
1 options
Catalog No. S07957 Supplier 3B Scientific™ Supplier No. W44119P
Add to Cart
Add to Cart
Request a Quote
Includes
Storage case for simulator, one remote control and one SmartScope with single- and dual-user headpieces
Simulator to train auscultation of various heart and lung sounds as in real patient.
  • Select various conditions by wireless remote control
  • Student identifies correct auscultation sites by palpating heart and lung sounds and then hears it by using special Stethoscope (SmartScope™)
  • Six heart and two lung sites on anterior and 16 lung sites on posterior
  • Single remote control to operate multiple sets of SmartScopes and manikins simultaneously
  • Remote control works over range of up to 30m
  • Includes three AAA batteries
The following sounds can be auscultated:
Heart sounds:
  • Normal
  • Aortic regurgitation
  • Pulmonary stenosis
  • Mitral stenosis
  • Holosystolic
  • Mid-systolic
  • S3 Gallop
  • S4 Gallop
  • Systolic click
  • Atrial septal defect
  • PDA
  • VSD
Lung sounds:
  • Normal tracheal
  • Normal vesicular
  • Wheezes
  • Mono wheeze
  • Fine crackle
  • Coarse crackle
  • Ronchi crackle
  • Stridor
  • Cavernous
  • Bronchovesicular
  • Bronchial
  • Pulmonary edema
  • Infant
  • Friction rub
  • Egophony
  • Pectoriloquy
Specifications
Product Type Auscultation Trainer Package with Smartscope and Amplifiers
Includes Storage case for simulator, one remote control and one SmartScope with single- and dual-user headpieces