Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Basic Hall Effect Apparatus
SDPSCI_ED

Catalog No. S08153 Shop All 3B Scientific Products
Change view
Click to view available options
Product Type:
Basic hall effect apparatus
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S08153 Basic hall effect apparatus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08153 Supplier 3B Scientific™ Supplier No. U8487000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Integrated, adjustable constant current source to provide current through sample
  • Measuring amplifier with offset compensation for Hall voltages and heating to raise crystal to as high as 170°C
  • Attachment for assembling apparatus on U-shaped core of transformer assembly kit
  • Two connecting leads with 8-pin miniDIN plugs

Provides contact, power supply and support to germanium crystal on circuit board in experiments on Hall effect or on conductivity.

  • Features temperature regulation and switchable display showing Hall voltage, sample current, sample voltage or temperature
  • Hall voltage and sample voltage can be tapped directly from front panel
  • Three equivalent voltage outputs for Hall voltage, sample current and sample temperature can be measured from side
Experiment topics:
  • Extrinsic conductivity
  • Intrinsic conductivity
  • Mobility of electrons and holes
  • Drift velocity of charge carriers
  • Carrier concentration
  • Band separation

  • Outputs for equivalent voltages: 4mm safety sockets
  • Power supply: 12VAC, 3A via 4-mm sockets
  • Dimensions: ∽18 x 11 x 5cm3
  • Weight: ∽0.5kg

Specifications

Product Type Basic hall effect apparatus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.