Learn More
3B Scientific™ Catheterization Trainers
SDPSCI_ED
Catalog No. S07976 Shop All 3B Scientific Products
Change view
Click to view available options
Product Type:
Trainer, Female Catheterization
Trainer, Male and Female Catheterization Set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S07976 Trainer, Male and Female Catheterization Set
S07974 Trainer, Female Catheterization
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S07976 Supplier 3B Scientific™ Supplier No. W44789SET
Add to Cart
Add to Cart
Material gives realistic feel and flexibility.
  • For training of urinary catheterization
  • Utilizes 16 FR catheter and can be used to demonstrate balloon inflation procedures
  • Fluid reservoir allows for urine output upon successful completion of inertion procedure
  • Instruction for to teach sterile technique and insertion and removal of catheter, as well as practice preparing patient for procedure
Specifications
Dimensions (L x W x H) 13.0 x 13.0 x 12.0 in.
For Use With (Application) Ideal for urinary catheterization
Weight (English) 6.5 lb.
Weight (Metric) 2.9 kg
Product Type Trainer, Male and Female Catheterization Set