Learn More
3B Scientific™ Deluxe Occluded Artery Model
SDPSCI_ED
Catalog No. S12790 Shop All 3B Scientific Products
Change view
Click to view available options
Deluxe Occluded Artery Model
Catalog No.
S12790 Deluxe Occluded Artery Model
1 options
Catalog No. S12790 Supplier 3B Scientific™ Supplier No. W43206D
Add to Cart
Add to Cart
Demonstrates benefits of healthy cholesterol level and deadly effects of high cholesterol.
  • Made of durable plastic
  • Two tubes represent normal artery and artery constricted by plaque
  • Blood in healthy artery flows swiftly to top but blood in occluded artery slows and even stops completely when model is turned over
Specifications
Product Type Model, Occluded Artery, Deluxe