Learn More
3B Scientific™ Electromagnetic Experiment Set
SDPSCI_ED
Catalog No. S05543 Shop All 3B Scientific Products
Change view
Click to view available options
Weight (Metric):
0.82 kg
Catalog No. Weight (Metric)
S05543 0.82 kg
1 options
Catalog No. S05543 Supplier 3B Scientific™ Supplier No. U10371
Add to Cart
Add to Cart
The Electromagnetic Experiment Set offers a concise set-up for experiments in advanced topics of electricity and magnetism such as induction and paramagnetism.
  • Consists of a stable, anodized-aluminum tripod with pre-defined magnet positions and accessory mountings
  • Deflection of the conductor swing can be adjusted in steps of 0, 15, 30 and 45mm for current balance experiments
  • All accessories can be fasted to the stand for storage
  • Scope of delivery: 1 Aluminum tripod, anodized, 1 Conductor swing with 4-mm safety jacks, 2 Waltenhofen pendulums (solid and slotted), 1 Glass rod and polyester thread with hook, 1 Aluminum rod and polyester thread with hook, 1 Knurled screw, 1 Allen Wrench
In-depth look at these experiments topics:
  • Behavior of a current-carrying conductor in a magnetic field
  • Induced eddy currents
  • Diamagnetism and paramagnetism
Specifications
Product Type Electromagnetic Experiment Set
Includes Aluminum tripod, anodized, 2 waltenhofen pendulums, Glass rod and Polyester thread with hook, 1 Aluminum rod and Polyester thread with hook, Knurled screw, 1 allen wrench
Weight (Metric) 0.82 kg
Weight (English) 1.81 lb.