Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Measuring without Instruments
SDPSCI_ED

Catalog No. S05515 Shop All 3B Scientific Products
Change view
Click to view available options
For student experiments at pre-secondary level
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S05515 For student experiments at pre-secondary level
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05515 Supplier 3B Scientific™ Supplier No. U10972

missing translation for 'validMainframeMsg'

Includes

1 Plug-in horn, 1 Plug-in incandescent lamp, 1 Plug-in motor, 1 Set of 5 rotary discs including holder

Supplement to the photovoltaic equipment set for student experiments at pre-secondary level.

Experiment Topics:

  • Electrical loads
  • Series and parallel connection of incandescent lamps
  • Direction of rotation and velocity of an electric motor
  • Brightness
  • Diffuse radiation
  • Albedo radiation
  • Rotating discs
  • Colour mixing
  • Colour illusions

Specifications

Product Type Kit, Measurement
For Use With (Application) For student experiments at pre secondary level
For Use With (Equipment) Photovoltaic equipment set
Includes 1 Plug in horn, 1 plug in incandescent lamp, 1 plug in motor, 1 set of 5 rotary discs including holder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.