Learn More
3B Scientific™ Occluded Artery Model
SDPSCI_ED
Catalog No. S12789 Shop All 3B Scientific Products
Change view
Click to view available options
Product Type:
Occluded Artery Model
Catalog No. Product Type
S12789 Occluded Artery Model
1 options
Catalog No. S12789 Supplier 3B Scientific™ Supplier No. W43206
Add to Cart
Add to Cart
Demonstrates deadly effects of high cholesterol.
  • Tube represents artery constricted by plaque, which forms on artery walls when there is excess cholesterol in the blood
  • Blood in artery slows or stops completely when model is turned over
  • Explains importance of keeping cholesterol under control
Specifications
Product Type Occluded Artery Model