Learn More
3B Scientific™ ORTHObones Human Sacrum Model
SDPSCI_ED
Catalog No. S07937 Shop All 3B Scientific Products
Change view
Click to view available options
Human sacrum model
Catalog No.
S07937 Human sacrum model
1 options
Catalog No. S07937 Supplier 3B Scientific™ Supplier No. W19124
Resembles human bone in terms of rigidity, density and lifelike structure
  • Used by surgeons, orthopaedists and medical engineers
  • Demonstrates corrective procedures used on sacral fractures or implanting and displaying medical devices
  • Realistic biochemical construction to simulate two distinct layers of real human bones; hard outer cortical shell and softer inner cancellous bone
Specifications
Dimensions (L x W x H) 4.3 x 3.5 x 2.4 in.
For Use With (Application) Demonstrate corrective procedures used on sacral fractures or implantation of medical devices
Weight (English) 0.55 lb.