Learn More
3B Scientific™ ORTHObones Human Tibia Models
SDPSCI_ED
Catalog No. S07935 Shop All 3B Scientific Products
Change view
Click to view available options
Product Type:
Model, Tibia, Right
Catalog No. Product Type
S07935 Model, Tibia, Right
1 options
Catalog No. S07935 Supplier 3B Scientific™ Supplier No. W19122
Resembles human bone in terms of form and characteristics.
  • Used by surgeons, orthopaedists and medical engineers
  • For practice of correcting leg fractures and demonstrate use of medical devices
  • Realistic construction to simulate two distinct layers of real human bones; hard outer cortical shell and softer inner cancellous bone
Specifications
Dimensions (L x W x H) 39 x 81 x 7 cm (15.4 x 31.9 x 2.8 in.)
For Use With (Application) Ideal for surgeons to practice correcting leg fractures, or for medical engineers to demonstrate the use of medical devices
Weight (English) 0.77 lb.
Weight (Metric) 0.35 kg
Product Type Model, Tibia, Right