Learn More
3B Scientific™ Pelvis Skeleton - includes 3B Smart Anatomy
GREENER_CHOICESDPSCI_ED
Catalog No. S171124 Shop All 3B Scientific Products
Change view
Click to view available options
Product Type:
Anatomical and Cellular Model, Female Pelvis Skeleton
Anatomical and Cellular Model, Male Pelvis Skeleton
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S171124 Anatomical and Cellular Model, Female Pelvis Skeleton
S171123 Anatomical and Cellular Model, Male Pelvis Skeleton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S171124 Supplier 3B Scientific™ Supplier No. A61
Add to Cart
Add to Cart
Illustrate the difference between the male and female pelvis.
3B Smart Anatomy is now included with any 3B Scientific anatomy model.
Every original 3B Scientific anatomy model now includes these additional FREE features:
  • Free access to the anatomy course 3B Smart Anatomy, hosted inside the award-winning Complete Anatomy app by 3D4Medical
  • The 3B Smart Anatomy course includes 23 digital anatomy lectures, 117 different virtual anatomy models and 39 anatomy quizzes to test your knowledge
  • FREE warranty upgrade from 3 to 5 years with every product registration
TIPP: you will also receive access to a free 3-day trial to all premium features of the Complete Anatomy app when you sign up for your 3B Smart Anatomy course.
To unlock these benefits, simply scan the label located on your model and register online. All 3B Smart Anatomy features are completely free of charge.
Both nondissectible models consist of two innominate bones, sacrum with coccyx and two lumbar vertebrae. Female model has a flexible symphysis.
Specifications
Includes Two innominate bones, sacrum with coccyx, two lumbar vertebrae, Flexible symphysis
Product Type Anatomical and Cellular Model, Female Pelvis Skeleton