Learn More
3B Scientific™ RNA 12-Base Kit
SDPSCI_ED
Catalog No. S68291A Shop All 3B Scientific Products
Change view
Click to view available options
Product Type:
RNA 12 Base Kit
Catalog No. Product Type
S68291A RNA 12 Base Kit
1 options
Catalog No. S68291A Supplier 3B Scientific™ Supplier No. W19761
Add to Cart
Add to Cart
Includes
  • 3 Uracil (light blue)
  • 3 Adenine (blue)
  • 3 Guanine (green)
  • 3 Cytosine (yellow)
  • 12 Ribose (red)
  • 12 Phosphate (purple)
Provides hands-on investigation into protein synthesis
Easily assemble this single strand molecule which consists of the four bases, as in DNA, and Uracil. Contains enough material to make all 64 codons.
Specifications
For Use With (Application) Provides hands-on investigation into protein synthesis
Includes 3 Uracil (light blue), 3 Adenine (blue), 3 Guanine (green), 3 Cytosine (yellow), 12 Ribose (red), 12 Phosphate (purple)
Product Type RNA 12 Base Kit