Learn More
3B Scientific™ Skin Cancer Model - includes 3B Smart Anatomy
GREENER_CHOICESDPSCI_ED
Catalog No. S90793 Shop All 3B Scientific Products
Change view
Click to view available options
Skin Cancer Model, Enlarged 8X
Catalog No.
S90793 Skin Cancer Model, Enlarged 8X
1 options
Catalog No. S90793 Supplier 3B Scientific™ Supplier No. J15
Add to Cart
Add to Cart
Vividly displays six different stages of the malignant melanoma.
3B Smart Anatomy is now included with any 3B Scientific anatomy model.
Every original 3B Scientific anatomy model now includes these additional FREE features:
  • Free access to the anatomy course 3B Smart Anatomy, hosted inside the award-winning Complete Anatomy app by 3D4Medical
  • The 3B Smart Anatomy course includes 23 digital anatomy lectures, 117 different virtual anatomy models and 39 anatomy quizzes to test your knowledge
  • FREE warranty upgrade from 3 to 5 years with every product registration
TIPP: you will also receive access to a free 3-day trial to all premium features of the Complete Anatomy app when you sign up for your 3B Smart Anatomy course.
To unlock these benefits, simply scan the label located on your model and register online. All 3B Smart Anatomy features are completely free of charge.
Enlarged eight times, this model shows great detail.
  • Sides show the various levels of invasion into the skin layers according to Clark (I-V) and the tumor thickness according to Breslow (in mm)
  • Top view shows the individual stages of externally visible skin changes, allowing for an assessment according to the ABCDE criteria
  • Five original color illustrations on the base show various types of malignant melanomas
  • Dimensions: 5.6 x 4 x 4.6 in. (14 x 10 x 11.5cm)
Specifications
For Use With (Application) Shows levels of invasion into skin layers, top shows individual stages of external skin changes, allows assessment using 'ABCDE' criteria
Includes Skin layers
Mounting Base
Weight (English) 0.44 lb.
Weight (Metric) 0.22 kg
Product Type Model, Skin Cancer