Learn More
3B Scientific™ Skull with Teeth for Extraction
SDPSCI_ED
Catalog No. S06813 Shop All 3B Scientific Products
Change view
Click to view available options
Skull with teeth for extraction
Catalog No.
S06813 Skull with teeth for extraction
1 options
Catalog No. S06813 Supplier 3B Scientific™ Supplier No. W10532
Add to Cart
Add to Cart
Ideal for dental and medical students.
  • Four-part model
  • Teeth can be extracted with their fully formed roots and replaced individually
  • Bone flap on right mandible can be opened to observe dental roots, spongiosa, nerve canal and an impacted wisdom tooth
  • Dimensions: 8.7 x 5.3 x 6.7 in. (22 x 13.5 x 17cm)
  • Weighs 1.76 lb. (0.8kg)
Specifications
Product Type Skull with Teeth for Extraction
For Use With (Application) Teaching and Learning of Human Anatomy, with a Special Focus on Teeth, Ideal for Dental and Medical Students
Weight (English) 1.76 lb.
Weight (Metric) 0.8 kg