Learn More
3B Scientific™ Spectrometry Supplementary Set
SDPSCI_ED
Catalog No. S05519 Shop All 3B Scientific Products
Change view
Click to view available options
Includes:
1 Spectrometer with SMA connectors, 1 reference light source with SMA connectors, Five 2m Fibre optic cables with SMA plugs, one 10m Fibre optic cable with SMA plugs, 5 SMA or SMA connectors
Catalog No. Includes
S05519 1 Spectrometer with SMA connectors, 1 reference light source with SMA connectors, Five 2m Fibre optic cables with SMA plugs, one 10m Fibre optic cable with SMA plugs, 5 SMA or SMA connectors
1 options
Catalog No. S05519 Supplier 3B Scientific™ Supplier No. U22056
Add to Cart
Add to Cart
Includes
One spectrometer with SMA connectors, one reference light source with SMA connectors, five 2m fibre optic cables with SMA plugs, one 10m fibre optic cable with SMA plugs, five SMA/SMA connectors
The 3B Scientific Spectrometry Supplementary Set is used for investigating spectrometry of transmitted signals and measurement of absorption losses.
  • Supplement to the “Optical telecommunications” equipment set
Specifications
Type Spectrometry Supplementary Set
Includes 1 Spectrometer with SMA connectors, 1 reference light source with SMA connectors, Five 2m Fibre optic cables with SMA plugs, one 10m Fibre optic cable with SMA plugs, 5 SMA or SMA connectors
For Use With (Application) For investigating spectrometry of transmitted signals, measurement of absorption losses
Weight (English) 2.65 lb.
Weight (Metric) 1.2 kg