Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Student Heat Experiments Set
SDPSCI_ED

Catalog No. S96475 Shop All 3B Scientific Products
Change view
Click to view available options
Class Size:
10 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S96475 10 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96475 Supplier 3B Scientific™ Supplier No. U60040
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

CD with experiment instructions; 100mL conical flask; bimetallic strip; 4g glycerine; concave mirror mounted on stem; 200mm aluminum rod; thermometer, –10 to +100°C; glass tube with 90° bend; rubber stopper, 25 x 24/19 mm dia.; spirit burner; and ceramic mat

Requires

3B Scientific Student Experiment Basic Set

Set of apparatus for carrying out 10 basic student experiments on heat

For use with 3B Scientific Student Experiment Basic Set.

  • Packed in a robust plastic box with foam inserts moulded to the shapes of the items and a transparent lid

Specifications

For Use With (Application) Set of apparatus for carrying out 10 basic student experiments on heat
Includes 1 conical flask, 100ml, 1 bimetallic strip, 4g glycerine, 1 concave mirror mounted on stem, 1 aluminum rod 200mm, 1 thermometer, -10°c to 100°c, 1 glass tube with 90° bend, 1 rubber stopper, 25 x 1.26m
Requires 3B Scientific student experiment basic set
Weight (English) 4.3 lb.
Weight (Metric) 1.95 kg
Class Size 10 Students
Product Type Student Kit - Heat Experiment Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.